Assay Details
| Gene Symbol: NR1D1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005584071.2 | XP_005584128.1 | 1 - 2 | 659 | 61 | |
| GenBank mRNA | AB173702.1 | - | 1 - 2 | 571 | 61 | |
More Information
Set Membership: |
|

NR1D1
Genomic Map