Assay Details
| Gene Symbol: GNA13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005584721.2 | XP_005584778.1 | 2 - 3 | 795 | 78 | |
| XM_015438721.1 | XP_015294207.1 | 2 - 3 | 765 | 78 | ||
More Information
Set Membership: |
|

GNA13
Genomic Map