Assay Details
| Gene Symbol: SIGLEC11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005596066.2 | XP_005596123.2 | 1 - 2 | 220 | 91 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005596066.2 transcripts.
Set Membership: |
|

SIGLEC11
Genomic Map