Assay Details
| Gene Symbol: SLC25A44 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005541471.2 | XP_005541528.1 | 3 - 4 | 1079 | 76 | |
| XM_005541472.2 | XP_005541529.1 | 4 - 5 | 1162 | 76 | ||
More Information
Set Membership: |
|

SLC25A44
Genomic Map