Assay Details
| Gene Symbol: UTS2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005545397.2 | XP_005545454.1 | 2 - 3 | 312 | 77 | |
| XM_015444924.1 | XP_015300410.1 | 3 - 4 | 472 | 77 | ||
| XM_015444925.1 | XP_015300411.1 | 1 - 2 | 309 | 77 | ||
| XM_015444926.1 | XP_015300412.1 | 2 - 3 | 312 | 77 | ||
More Information
Set Membership: |
|

UTS2B
Genomic Map