Assay Details
| Gene Symbol: LSMEM2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005547206.2 | XP_005547263.1 | 1 - 2 | 1167 | 73 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

LSMEM2| Gene Symbol: LSMEM2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005547206.2 | XP_005547263.1 | 1 - 2 | 1167 | 73 | |