Assay Details
| Gene Symbol: LSMEM2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005547206.2 | XP_005547263.1 | 3 - 4 | 1462 | 82 | |
| XM_005547207.2 | XP_005547264.1 | 3 - 4 | 1542 | 82 | ||
| XM_015445937.1 | XP_015301423.1 | 3 - 4 | 471 | 82 | ||
More Information
Set Membership: |
|

LSMEM2
Genomic Map