Assay Details
| Gene Symbol: LOC102123502 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015447058.1 | XP_015302544.1 | 5 - 6 | 1895 | 70 | |
| XM_015447059.1 | XP_015302545.1 | 5 - 6 | 780 | 70 | ||
| XM_015447060.1 | XP_015302546.1 | 5 - 6 | 701 | 70 | ||
| XM_015447061.1 | XP_015302547.1 | 5 - 6 | 1014 | 70 | ||
More Information
Set Membership: |
|

LOC102123502
Genomic Map