Assay Details
| Gene Symbol: LOC102123502 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015447058.1 | XP_015302544.1 | 6 - 7 | 1980 | 80 | |
| XM_015447059.1 | XP_015302545.1 | 6 - 7 | 865 | 80 | ||
| XM_015447060.1 | XP_015302546.1 | 6 - 7 | 786 | 80 | ||
| XM_015447061.1 | XP_015302547.1 | 6 - 7 | 1099 | 80 | ||
| XM_015447062.1 | XP_015302548.1 | 5 - 6 | 1806 | 80 | ||
| XM_015447063.1 | XP_015302549.1 | 4 - 5 | 1737 | 80 | ||

LOC102123502
Genomic Map