Assay Details
| Gene Symbol: LOC102123502 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015447058.1 | XP_015302544.1 | 7 - 8 | 2103 | 94 | |
| XM_015447059.1 | XP_015302545.1 | 7 - 8 | 988 | 94 | ||
| XM_015447060.1 | XP_015302546.1 | 7 - 8 | 909 | 94 | ||
| XM_015447061.1 | XP_015302547.1 | 7 - 8 | 1222 | 94 | ||
| XM_015447062.1 | XP_015302548.1 | 6 - 7 | 1929 | 94 | ||
| XM_015447063.1 | XP_015302549.1 | 5 - 6 | 1860 | 94 | ||

LOC102123502
Genomic Map