Assay Details
| Gene Symbol: LOC102123502 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015447058.1 | XP_015302544.1 | 8 - 9 | 2276 | 63 | |
| XM_015447059.1 | XP_015302545.1 | 8 - 9 | 1161 | 63 | ||
| XM_015447060.1 | XP_015302546.1 | 8 - 9 | 1082 | 63 | ||
| XM_015447061.1 | XP_015302547.1 | 8 - 9 | 1395 | 63 | ||
| XM_015447062.1 | XP_015302548.1 | 7 - 8 | 2102 | 63 | ||
| XM_015447063.1 | XP_015302549.1 | 6 - 7 | 2033 | 63 | ||
More Information
Set Membership: |
|

LOC102123502
Genomic Map