Assay Details
| Gene Symbol: C5H4orf48 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005554303.2 | XP_005554360.2 | 3 - 4 | 339 | 77 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

C5H4orf48| Gene Symbol: C5H4orf48 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005554303.2 | XP_005554360.2 | 3 - 4 | 339 | 77 | |
Set Membership: |
|