Assay Details
| Gene Symbol: LOC102140933 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005558195.2 | XP_005558252.1 | 3 - 4 | 1154 | 127 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC102140933| Gene Symbol: LOC102140933 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005558195.2 | XP_005558252.1 | 3 - 4 | 1154 | 127 | |
Set Membership: |
|