Assay Details
Gene Symbol: LOC102143735 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015455822.1 | XP_015311308.1 | 23 - 24 | 7883 | 91 | |
XM_015455823.1 | XP_015311309.1 | 24 - 25 | 7051 | 91 | ||
XM_015455824.1 | XP_015311310.1 | 24 - 25 | 4928 | 91 | ||
XM_015455825.1 | XP_015311311.1 | 23 - 24 | 7883 | 91 | ||
XM_015455826.1 | XP_015311312.1 | 23 - 24 | 7883 | 91 | ||
XM_015455828.1 | XP_015311314.1 | 23 - 24 | 7883 | 91 | ||
XM_015455829.1 | XP_015311315.1 | 23 - 24 | 7883 | 91 | ||
XM_015455830.1 | XP_015311316.1 | 23 - 24 | 7880 | 91 | ||
XM_015455831.1 | XP_015311317.1 | 23 - 24 | 4381 | 91 | ||
XM_015455832.1 | XP_015311318.1 | 22 - 23 | 4075 | 91 | ||
XM_015455833.1 | XP_015311319.1 | 22 - 23 | 3357 | 91 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
