Assay Details
| Gene Symbol: LOC102143735 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015455822.1 | XP_015311308.1 | 6 - 7 | 5186 | 98 | |
| XM_015455823.1 | XP_015311309.1 | 7 - 8 | 4354 | 98 | ||
| XM_015455824.1 | XP_015311310.1 | 7 - 8 | 2231 | 98 | ||
| XM_015455825.1 | XP_015311311.1 | 6 - 7 | 5186 | 98 | ||
| XM_015455826.1 | XP_015311312.1 | 6 - 7 | 5186 | 98 | ||
| XM_015455828.1 | XP_015311314.1 | 6 - 7 | 5186 | 98 | ||
| XM_015455829.1 | XP_015311315.1 | 6 - 7 | 5186 | 98 | ||
| XM_015455831.1 | XP_015311317.1 | 6 - 7 | 1684 | 98 | ||
| XM_015455832.1 | XP_015311318.1 | 5 - 6 | 1378 | 98 | ||
| XM_015455833.1 | XP_015311319.1 | 5 - 6 | 660 | 98 | ||

LOC102143735
Genomic Map