Assay Details
| Gene Symbol: DYNLRB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005568768.2 | XP_005568825.1 | 3 - 4 | 359 | 95 | |
| XM_015457957.1 | XP_015313443.1 | 3 - 4 | 305 | 95 | ||
More Information
Set Membership: |
|

DYNLRB1
Genomic Map