Assay Details
| Gene Symbol: AURKA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005569391.2 | XP_005569448.1 | 7 - 8 | 992 | 118 | |
| XM_005569392.1 | XP_005569449.1 | 7 - 8 | 1002 | 118 | ||
| XM_005569394.2 | XP_005569451.1 | 7 - 8 | 1006 | 118 | ||
| XM_015429987.1 | XP_015285473.1 | 7 - 8 | 997 | 118 | ||
| XM_015429988.1 | XP_015285474.1 | 7 - 8 | 884 | 118 | ||
| GenBank mRNA | AB168134.1 | - | 7 - 8 | 925 | 118 | |
More Information
Set Membership: |
|

AURKA
Genomic Map