Assay Details
| Gene Symbol: PCBP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005575665.2 | XP_005575722.1 | 1 | 825 | 97 | |
| GenBank mRNA | AB174678.1 | - | 1 | 564 | 97 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

PCBP1| Gene Symbol: PCBP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005575665.2 | XP_005575722.1 | 1 | 825 | 97 | |
| GenBank mRNA | AB174678.1 | - | 1 | 564 | 97 | |
Set Membership: |
|