Assay Details
| Gene Symbol: GALK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005584980.2 | XP_005585037.1 | 5 - 6 | 1370 | 55 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

GALK1| Gene Symbol: GALK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005584980.2 | XP_005585037.1 | 5 - 6 | 1370 | 55 | |
Set Membership: |
|