Assay Details
| Gene Symbol: ANKRD30B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005587282.2 | XP_005587339.1 | 10 - 11 | 1139 | 91 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ANKRD30B| Gene Symbol: ANKRD30B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005587282.2 | XP_005587339.1 | 10 - 11 | 1139 | 91 | |
Set Membership: |
|