Assay Details
| Gene Symbol: ELOF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005588062.2 | XP_005588119.1 | 2 - 3 | 317 | 67 | |
| XM_005588063.2 | XP_005588120.1 | 2 - 3 | 343 | 67 | ||
More Information
Set Membership: |
|

ELOF1
Genomic Map