Assay Details
| Gene Symbol: CYP4F11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001319493.1 | NP_001306422.1 | 10 - 11 | 1403 | 84 | |
| XM_005588306.2 | XP_005588363.1 | 11 - 12 | 1367 | 84 | ||
| XM_005588307.2 | XP_005588364.1 | 11 - 12 | 1381 | 84 | ||
| GenBank mRNA | DQ074800.1 | - | 10 - 11 | 1403 | 84 | |
More Information
Set Membership: |
|

CYP4F11
Genomic Map