Assay Details
| Gene Symbol: CYP4F11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001319493.1 | NP_001306422.1 | 8 - 9 | 1201 | 82 | |
| XM_005588306.2 | XP_005588363.1 | 9 - 10 | 1165 | 82 | ||
| XM_005588307.2 | XP_005588364.1 | 9 - 10 | 1179 | 82 | ||
| GenBank mRNA | DQ074800.1 | - | 8 - 9 | 1201 | 82 | |
More Information
Set Membership: |
|

CYP4F11
Genomic Map