Assay Details
| Gene Symbol: MRPS21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005542014.2 | XP_005542071.1 | 2 - 3 | 210 | 97 | |
| XM_005542015.2 | XP_005542072.1 | 2 - 3 | 202 | 97 | ||
More Information
Set Membership: |
|

MRPS21
Genomic Map