Assay Details
| Gene Symbol: TTC4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005543273.2 | XP_005543330.1 | 4 - 5 | 547 | 69 | |
| XM_005543274.2 | XP_005543331.1 | 4 - 5 | 547 | 69 | ||
More Information
Set Membership: |
|

TTC4
Genomic Map