Assay Details
| Gene Symbol: ZNF107 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005549452.2 | XP_005549509.1 | 1 - 2 | 657 | 89 | |
| XM_005549453.2 | XP_005549510.1 | 1 - 2 | 661 | 89 | ||
More Information
Set Membership: |
|

ZNF107
Genomic Map