Assay Details
| Gene Symbol: LOC102132272 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005552112.2 | XP_005552169.1 | 1 - 2 | 442 | 94 | |
| XM_005552113.2 | XP_005552170.1 | 1 - 2 | 443 | 94 | ||
 Back To Top
 Back To TopMore Information
| Set Membership: |  Amplicon length between 86 and 100  3' Most  Recommended Assay  Probe spans exons | 
Panther Classification:
 Back To Top
 Back To Top
 LOC102132272
LOC102132272 Genomic Map
Genomic Map Back To Top
 Back To Top