Assay Details
| Gene Symbol: FAM153A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005558673.2 | XP_005558730.1 | 6 - 7 | 402 | 83 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

FAM153A| Gene Symbol: FAM153A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005558673.2 | XP_005558730.1 | 6 - 7 | 402 | 83 | |