Assay Details
| Gene Symbol: LOC102143022 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015452577.1 | XP_015308063.1 | 8 - 9 | 698 | 64 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC102143022| Gene Symbol: LOC102143022 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015452577.1 | XP_015308063.1 | 8 - 9 | 698 | 64 | |
Set Membership: |
|