Assay Details
| Gene Symbol: NRL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015453620.1 | XP_015309106.1 | 1 - 2 | 1503 | 62 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015453620.1 transcripts.
Set Membership: |
|

NRL
Genomic Map