Assay Details
| Gene Symbol: C7H14orf180 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005562337.2 | XP_005562394.1 | 6 - 7 | 900 | 114 | |
| XM_005562339.2 | XP_005562396.1 | 6 - 7 | 1930 | 114 | ||
| XM_005562340.2 | XP_005562397.1 | 6 - 7 | 1908 | 114 | ||
| XM_005562341.2 | XP_005562398.1 | 4 - 5 | 773 | 114 | ||
| XM_015454252.1 | XP_015309738.1 | 6 - 7 | 842 | 114 | ||
| XM_015454253.1 | XP_015309739.1 | 4 - 5 | 1809 | 114 | ||
More Information
Set Membership: |
|

C7H14orf180
Genomic Map