Assay Details
| Gene Symbol: LOC102137707 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015457107.1 | XP_015312593.1 | 9 - 10 | 823 | 91 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC102137707| Gene Symbol: LOC102137707 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015457107.1 | XP_015312593.1 | 9 - 10 | 823 | 91 | |
Set Membership: |
|