Assay Details
| Gene Symbol: CST5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005568164.2 | XP_005568221.1 | 1 - 2 | 343 | 98 | |
| XM_015457730.1 | XP_015313216.1 | 2 - 3 | 659 | 98 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

CST5| Gene Symbol: CST5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005568164.2 | XP_005568221.1 | 1 - 2 | 343 | 98 | |
| XM_015457730.1 | XP_015313216.1 | 2 - 3 | 659 | 98 | ||