Assay Details
| Gene Symbol: LOC102132378 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005570065.2 | XP_005570122.1 | 24 - 25 | 4880 | 73 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC102132378| Gene Symbol: LOC102132378 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005570065.2 | XP_005570122.1 | 24 - 25 | 4880 | 73 | |
Set Membership: |
|