Assay Details
| Gene Symbol: LOC102132378 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005570065.2 | XP_005570122.1 | 28 - 29 | 5367 | 80 | |
| XM_015430345.1 | XP_015285831.1 | 27 - 28 | 5190 | 80 | ||
More Information
Set Membership: |
|

LOC102132378
Genomic Map