Assay Details
| Gene Symbol: LOC102132378 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005570065.2 | XP_005570122.1 | 30 - 31 | 5809 | 92 | |
| XM_015430345.1 | XP_015285831.1 | 29 - 30 | 5632 | 92 | ||
More Information
Set Membership: |
|

LOC102132378
Genomic Map