Assay Details
| Gene Symbol: LOC102132378 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005570065.2 | XP_005570122.1 | 9 - 10 | 2835 | 64 | |
| XM_015430345.1 | XP_015285831.1 | 9 - 10 | 2835 | 64 | ||
More Information
Set Membership: |
|

LOC102132378
Genomic Map