Assay Details
| Gene Symbol: LOC102136393 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001319494.1 | NP_001306423.1 | 11 - 12 | 1035 | 67 | |
| XM_015437764.1 | XP_015293250.1 | 11 - 12 | 2170 | 67 | ||
| XM_015437765.1 | XP_015293251.1 | 10 - 11 | 1005 | 67 | ||
| XM_015437766.1 | XP_015293252.1 | 11 - 12 | 1119 | 67 | ||
| GenBank mRNA | KP731383.1 | - | 11 - 12 | 1035 | 67 | |
More Information
Set Membership: |
|

LOC102136393
Genomic Map