Assay Details
| Gene Symbol: LOC102136393 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001319494.1 | NP_001306423.1 | 13 - 14 | 1178 | 81 | |
| XM_015437764.1 | XP_015293250.1 | 13 - 14 | 2313 | 81 | ||
| XM_015437765.1 | XP_015293251.1 | 12 - 13 | 1148 | 81 | ||
| XM_015437766.1 | XP_015293252.1 | 13 - 14 | 1262 | 81 | ||
| GenBank mRNA | KP731383.1 | - | 13 - 14 | 1178 | 81 | |
More Information
Set Membership: |
|

LOC102136393
Genomic Map