Assay Details
| Gene Symbol: LOC102136393 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001319494.1 | NP_001306423.1 | 15 - 16 | 1405 | 103 | |
| XM_015437764.1 | XP_015293250.1 | 15 - 16 | 2540 | 103 | ||
| XM_015437765.1 | XP_015293251.1 | 14 - 15 | 1375 | 103 | ||
| GenBank mRNA | KP731383.1 | - | 15 - 16 | 1405 | 103 | |
More Information
Set Membership: |
|

LOC102136393
Genomic Map