Assay Details
| Gene Symbol: TPM4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005588309.2 | XP_005588366.1 | 2 - 3 | 384 | 54 | |
| XM_005588310.2 | XP_005588367.1 | 2 - 3 | 387 | 54 | ||
| XM_005588311.2 | XP_005588368.1 | 2 - 3 | 395 | 54 | ||
More Information
Set Membership: |
|

TPM4
Genomic Map