Assay Details
| Gene Symbol: C16H17orf77 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005596052.2 | XP_005596109.1 | 1 | 1405 | 86 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

C16H17orf77| Gene Symbol: C16H17orf77 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005596052.2 | XP_005596109.1 | 1 | 1405 | 86 | |
Set Membership: |
|