Assay Details
| Gene Symbol: LOC102119850 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005595158.2 | XP_005595215.1 | 470 | 108 | ||
| XM_015444375.1 | XP_015299861.1 | 470 | 108 | |||
| XM_015444376.1 | XP_015299862.1 | 470 | 108 | |||
| XM_015444377.1 | XP_015299863.1 | 470 | 108 | |||
More Information
Set Membership: |
|

LOC102119850
Genomic Map