Assay Details
| Gene Symbol: LOC102121133 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015448906.1 | XP_015304392.1 | 7 - 8 | 1065 | 94 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC102121133| Gene Symbol: LOC102121133 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015448906.1 | XP_015304392.1 | 7 - 8 | 1065 | 94 | |
Set Membership: |
|