Assay Details
| Gene Symbol: LOC102146239 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005595955.2 | XP_005596012.2 | 6 - 7 | 527 | 85 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC102146239| Gene Symbol: LOC102146239 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005595955.2 | XP_005596012.2 | 6 - 7 | 527 | 85 | |
Set Membership: |
|