Assay Details
| Gene Symbol: LOC102144764 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005590337.2 | XP_005590394.1 | 1 - 2 | 35 | 77 | |
| GenBank mRNA | EU419108.1 | - | 1 - 2 | 35 | 77 | |
More Information
Set Membership: |
|

LOC102144764
Genomic Map