Assay Details
| Gene Symbol: RPS14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005558271.2 | XP_005558328.1 | 3 - 4 | 436 | 75 | |
| XM_005558272.2 | XP_005558329.1 | 3 - 4 | 574 | 75 | ||
More Information
Set Membership: |
|

RPS14
Genomic Map