Assay Details
| Gene Symbol: LOC102140229 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005590834.2 | XP_005590891.1 | 3 - 4 | 943 | 107 | |
| GenBank mRNA | EU341822.1 | - | 3 - 4 | 480 | 107 | |
More Information
Set Membership: |
|

LOC102140229
Genomic Map