Assay Details
| Gene Symbol: NIPSNAP3B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005581146.2 | XP_005581203.1 | 3 - 4 | 936 | 88 | |
| XM_015436906.1 | XP_015292392.1 | 2 - 3 | 824 | 88 | ||
More Information
Set Membership: |
|

NIPSNAP3B
Genomic Map