Assay Details
| Gene Symbol: LOC102119798 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005565310.2 | XP_005565367.2 | 3 | 337 | 90 | |
| XM_005565311.2 | XP_005565368.1 | 4 | 340 | 90 | ||
| XM_005565313.2 | XP_005565370.1 | 4 | 307 | 90 | ||
More Information
Set Membership: |
|

LOC102119798
Genomic Map